Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273603.2 | XP_015129089.1 | 3 - 4 | 460 | 142 | |
XM_015273607.2 | XP_015129093.1 | 3 - 4 | 336 | 142 | ||
XM_416834.5 | XP_416834.1 | 3 - 4 | 503 | 142 | ||
GenBank mRNA | BX929875.2 | - | 3 - 4 | 340 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |