Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935629.3 | XP_004935686.1 | 2 - 3 | 294 | 90 | |
XM_025149276.1 | XP_025005044.1 | 2 - 3 | 594 | 90 | ||
XM_419678.6 | XP_419678.4 | 2 - 3 | 593 | 90 | ||
GenBank mRNA | BX931430.2 | - | 2 - 3 | 98 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |