Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277810.1 | NP_001264739.1 | 1 - 2 | 42 | 111 | |
XM_015280010.2 | XP_015135496.1 | 1 - 2 | 99 | 111 | ||
XM_015280011.2 | XP_015135497.1 | 1 - 2 | 70 | 111 | ||
GenBank mRNA | BI065449.1 | - | 1 - 2 | 36 | 111 | |
BM489788.1 | - | 1 - 2 | 42 | 111 | ||
BX935001.2 | - | 1 - 2 | 35 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |