Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX935790.2 | - | 7 - 8 | 830 | 138 | |
BX935866.2 | - | 7 - 8 | 830 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |