Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422114.6 | XP_422114.4 | 3 - 4 | 1934 | 121 | |
GenBank mRNA | CR352522.1 | - | 1 - 2 | 878 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |