Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277567.1 | NP_001264496.1 | 3 - 4 | 385 | 138 | |
XM_015283652.2 | XP_015139138.1 | 3 - 4 | 707 | 138 | ||
XM_015283653.2 | XP_015139139.1 | 3 - 4 | 1225 | 138 | ||
GenBank mRNA | BU372764.1 | - | 3 - 4 | 355 | 138 | |
BU404580.1 | - | 3 - 4 | 385 | 138 | ||
BX931356.2 | - | 3 - 4 | 371 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |