Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271911.1 | NP_001258840.1 | 2 - 3 | 206 | 66 | |
NM_205356.2 | NP_990687.2 | 2 - 3 | 206 | 66 | ||
GenBank mRNA | BG641954.1 | - | 2 - 3 | 206 | 66 | |
BX935039.1 | - | 2 - 3 | 188 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |