Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX935521.1 | - | 2 - 3 | 70 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |