Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012932.2 | NP_001012950.2 | 10 - 11 | 1741 | 78 | |
XM_015298617.2 | XP_015154103.2 | 10 - 11 | 2004 | 78 | ||
XM_015298619.2 | XP_015154105.2 | 10 - 11 | 2002 | 78 | ||
XM_015298620.2 | XP_015154106.2 | 11 - 12 | 1926 | 78 | ||
XM_015298621.2 | XP_015154107.2 | 9 - 10 | 1927 | 78 | ||
XM_015298622.2 | XP_015154108.2 | 11 - 12 | 1923 | 78 | ||
XM_015298623.2 | XP_015154109.2 | 9 - 10 | 1924 | 78 | ||
GenBank mRNA | AJ720606.1 | - | 10 - 11 | 1741 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |