Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949167.3 | XP_004949224.3 | 6 - 7 | 1497 | 130 | |
XM_004949169.3 | XP_004949226.1 | 7 - 8 | 720 | 130 | ||
XM_004949170.3 | XP_004949227.1 | 6 - 7 | 730 | 130 | ||
XM_004949171.3 | XP_004949228.1 | 6 - 7 | 604 | 130 | ||
XM_004949172.1 | XP_004949229.1 | 6 - 7 | 552 | 130 | ||
XM_015280351.2 | XP_015135837.1 | 7 - 8 | 616 | 130 | ||
XM_025145268.1 | XP_025001036.1 | 6 - 7 | 735 | 130 | ||
XM_025145269.1 | XP_025001037.1 | 7 - 8 | 640 | 130 | ||
XM_424839.5 | XP_424839.2 | 6 - 7 | 544 | 130 | ||
GenBank mRNA | BX931137.1 | - | 6 - 7 | 523 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |