Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006321.1 | NP_001006321.1 | 2 - 3 | 194 | 84 | |
GenBank mRNA | AJ720714.1 | - | 2 - 3 | 194 | 84 | |
CR405813.1 | - | 2 - 3 | 174 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |