Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006350.2 | NP_001006350.1 | 2 - 3 | 334 | 138 | |
GenBank mRNA | AJ719425.1 | - | 2 - 3 | 336 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |