Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006394.1 | NP_001006394.1 | 2 - 3 | 209 | 93 | |
XM_015283363.2 | XP_015138849.1 | 2 - 3 | 212 | 93 | ||
XM_015283364.2 | XP_015138850.1 | 2 - 3 | 212 | 93 | ||
XM_015283365.2 | XP_015138851.1 | 2 - 3 | 212 | 93 | ||
XM_015283366.2 | XP_015138852.1 | 2 - 3 | 212 | 93 | ||
XM_015283367.2 | XP_015138853.1 | 2 - 3 | 209 | 93 | ||
XM_015283369.2 | XP_015138855.1 | 2 - 3 | 209 | 93 | ||
XM_015283370.2 | XP_015138856.1 | 2 - 3 | 209 | 93 | ||
XM_015283371.2 | XP_015138857.1 | 2 - 3 | 209 | 93 | ||
XM_025148639.1 | XP_025004407.1 | 2 - 3 | 212 | 93 | ||
XM_025148640.1 | XP_025004408.1 | 2 - 3 | 209 | 93 | ||
GenBank mRNA | AJ720271.1 | - | 2 - 3 | 209 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |