Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006460.1 | NP_001006460.1 | 3 - 4 | 340 | 84 | |
XM_015285898.2 | XP_015141384.1 | 3 - 4 | 404 | 84 | ||
XM_015285899.2 | XP_015141385.1 | 3 - 4 | 351 | 84 | ||
GenBank mRNA | AJ719815.1 | - | 3 - 4 | 340 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |