Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204606.1 | NP_989937.1 | 3 - 4 | 268 | 102 | |
GenBank mRNA | AF411083.1 | - | 3 - 4 | 268 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |