Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204806.2 | NP_990137.2 | 2 - 3 | 1190 | 138 | |
GenBank mRNA | AF106940.1 | - | 2 - 3 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |