Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204852.1 | NP_990183.1 | 7 - 8 | 965 | 92 | |
XM_015297601.2 | XP_015153087.1 | 7 - 8 | 1001 | 92 | ||
GenBank mRNA | AF097750.1 | - | 7 - 8 | 965 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |