Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204852.1 | NP_990183.1 | 8 - 9 | 1044 | 88 | |
XM_015297601.2 | XP_015153087.1 | 8 - 9 | 1080 | 88 | ||
GenBank mRNA | AF097750.1 | - | 8 - 9 | 1044 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |