Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205358.1 | NP_990689.2 | 4 - 5 | 744 | 92 | |
GenBank mRNA | L39880.1 | - | 4 - 5 | 744 | 92 | |
U41823.1 | - | 4 - 5 | 669 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |