Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205366.2 | NP_990697.2 | 3 - 4 | 443 | 148 | |
GenBank mRNA | BU355061.1 | - | 3 - 4 | 426 | 149 | |
U09350.1 | - | 3 - 4 | 419 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |