Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205417.1 | NP_990748.1 | 6 - 7 | 477 | 74 | |
GenBank mRNA | BX930062.2 | - | 486 | 74 | ||
U19926.1 | - | 6 - 7 | 477 | 74 | ||
X03832.1 | - | 191 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |