Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205479.3 | NP_990810.2 | 2 - 3 | 212 | 79 | |
GenBank mRNA | J02823.1 | - | 2 - 3 | 138 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |