Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205515.1 | NP_990846.1 | 471 | 92 | ||
GenBank mRNA | AJ719691.1 | - | 161 | 92 | ||
M25534.1 | - | 471 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus |