Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205528.1 | NP_990859.1 | 3 - 4 | 478 | 101 | |
GenBank mRNA | AJ719693.1 | - | 3 - 4 | 435 | 101 | |
J02912.1 | - | 3 - 4 | 478 | 101 | ||
M55660.1 | - | 3 - 4 | 489 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |