Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006375.1 | NP_001006375.1 | 1 - 2 | 169 | 87 | |
XM_015275890.2 | XP_015131376.1 | 5 - 6 | 2128 | 87 | ||
XM_015275891.2 | XP_015131377.1 | 5 - 6 | 2128 | 87 | ||
GenBank mRNA | AJ719521.1 | - | 1 - 2 | 169 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |