Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012561.1 | NP_001012579.1 | 3 - 4 | 603 | 61 | |
GenBank mRNA | AJ851471.1 | - | 3 - 4 | 603 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |