Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012699.1 | NP_001012717.1 | 7 - 8 | 959 | 74 | |
GenBank mRNA | AJ851448.1 | - | 7 - 8 | 959 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |