Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012788.1 | NP_001012806.1 | 9 - 10 | 1754 | 65 | |
GenBank mRNA | AJ719427.1 | - | 9 - 10 | 1781 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |