Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012833.1 | NP_001012851.1 | 3 - 4 | 480 | 113 | |
XM_015296384.2 | XP_015151870.2 | 3 - 4 | 131 | 113 | ||
XM_015296385.2 | XP_015151871.2 | 2 - 3 | 156 | 113 | ||
GenBank mRNA | AJ719437.1 | - | 3 - 4 | 472 | 113 | |
AJ851651.1 | - | 3 - 4 | 480 | 113 | ||
BX933801.1 | - | 3 - 4 | 218 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |