Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCCPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012875.1 | NP_001012893.1 | 7 - 8 | 877 | 67 | |
GenBank mRNA | AJ719381.1 | - | 7 - 8 | 877 | 67 | |
CR353828.1 | - | 7 - 8 | 842 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |