Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244738.1 | NP_001231667.1 | 6 - 7 | 1008 | 135 | |
NR_045197.1 | 6 - 7 | 1000 | 135 | |||
GenBank mRNA | AJ719535.1 | - | 6 - 7 | 954 | 135 | |
BX931412.1 | - | 4 - 5 | 622 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |