Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012997.1 | NP_001013015.1 | 2 - 3 | 235 | 103 | |
GenBank mRNA | BX931585.2 | - | 2 - 3 | 233 | 103 | |
BX931605.1 | - | 1 - 2 | 103 | 103 | ||
BX950478.1 | - | 2 - 3 | 235 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |