Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013394.1 | NP_001013412.1 | 9 - 10 | 1247 | 93 | |
GenBank mRNA | AJ719435.1 | - | 9 - 10 | 1247 | 93 | |
CR523416.1 | - | 8 - 9 | 952 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |