Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030872.2 | NP_001026043.1 | 3 - 4 | 558 | 68 | |
XM_015296869.2 | XP_015152355.1 | 3 - 4 | 761 | 68 | ||
XM_015296870.2 | XP_015152356.1 | 3 - 4 | 758 | 68 | ||
XM_015296871.2 | XP_015152357.1 | 3 - 4 | 699 | 68 | ||
GenBank mRNA | AJ720451.1 | - | 3 - 4 | 560 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |