Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030879.1 | NP_001026050.1 | 1 - 2 | 608 | 68 | |
XM_015296894.2 | XP_015152380.1 | 2 - 3 | 671 | 68 | ||
XM_015296895.2 | XP_015152381.1 | 2 - 3 | 609 | 68 | ||
GenBank mRNA | AJ720006.1 | - | 1 - 2 | 608 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |