Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031214.1 | NP_001026385.1 | 5 - 6 | 344 | 140 | |
GenBank mRNA | AJ720612.1 | - | 5 - 6 | 344 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |