Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001464.1 | NP_001001464.1 | 7 - 8 | 1560 | 96 | |
XM_015289929.2 | XP_015145415.1 | 8 - 9 | 1442 | 96 | ||
XM_015289937.2 | XP_015145423.1 | 8 - 9 | 1532 | 96 | ||
XM_015289940.2 | XP_015145426.1 | 8 - 9 | 2193 | 96 | ||
XM_015289942.2 | XP_015145428.1 | 8 - 9 | 1517 | 96 | ||
XM_015289947.2 | XP_015145433.1 | 8 - 9 | 1579 | 96 | ||
XM_015289951.2 | XP_015145437.1 | 7 - 8 | 1527 | 96 | ||
XM_015289959.2 | XP_015145445.1 | 6 - 7 | 1356 | 96 | ||
GenBank mRNA | AF163809.1 | - | 7 - 8 | 1560 | 96 | |
AF470455.1 | - | 7 - 8 | 1560 | 96 | ||
AJ007664.1 | - | 1 - 2 | 347 | 96 | ||
EU049891.1 | - | 5 - 6 | 1184 | 96 | ||
KT318735.1 | - | 5 - 6 | 1184 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |