Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001530.1 | NP_001001530.1 | 14 - 15 | 1990 | 69 | |
XM_015291385.2 | XP_015146871.1 | 14 - 15 | 1681 | 69 | ||
GenBank mRNA | AY463529.1 | - | 14 - 15 | 1990 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |