Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003797.1 | NP_001003797.1 | 4 - 5 | 1385 | 85 | |
XM_015293077.2 | XP_015148563.1 | 4 - 5 | 1494 | 85 | ||
XM_015293078.2 | XP_015148564.1 | 4 - 5 | 1350 | 85 | ||
XM_015293079.2 | XP_015148565.1 | 4 - 5 | 1458 | 85 | ||
XM_015293080.2 | XP_015148566.1 | 3 - 4 | 905 | 85 | ||
GenBank mRNA | X56930.1 | - | 1385 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |