Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004369.1 | NP_001004369.1 | 2 - 3 | 201 | 71 | |
XM_015278892.2 | XP_015134378.1 | 3 - 4 | 342 | 71 | ||
XM_015278893.2 | XP_015134379.1 | 2 - 3 | 428 | 71 | ||
GenBank mRNA | X62531.1 | - | 201 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |