Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204305.1 | NP_989636.1 | 4 - 5 | 292 | 87 | |
GenBank mRNA | AF047874.1 | - | 4 - 5 | 292 | 87 | |
J00850.1 | - | 2 - 3 | 169 | 87 | ||
JQ280469.1 | - | 2 - 3 | 177 | 87 | ||
K01458.1 | - | 4 - 5 | 291 | 87 | ||
V00407.1 | - | 3 - 4 | 264 | 87 | ||
X01578.1 | - | 239 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |