Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204305.1 | NP_989636.1 | 9 - 10 | 805 | 83 | |
GenBank mRNA | AF047874.1 | - | 9 - 10 | 805 | 83 | |
AY466166.1 | - | 2 - 3 | 164 | 83 | ||
J00850.1 | - | 7 - 8 | 682 | 83 | ||
K01458.1 | - | 9 - 10 | 804 | 83 | ||
V00406.1 | - | 160 | 83 | |||
V00407.1 | - | 8 - 9 | 777 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |