Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204849.1 | NP_990180.1 | 5 - 6 | 707 | 80 | |
GenBank mRNA | AF131224.1 | - | 5 - 6 | 707 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |