Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204944.2 | NP_990275.1 | 2 - 3 | 1137 | 82 | |
XM_015294136.2 | XP_015149622.1 | 2 - 3 | 1367 | 82 | ||
XM_015294137.2 | XP_015149623.1 | 2 - 3 | 1370 | 82 | ||
XM_025155049.1 | XP_025010817.1 | 3 - 4 | 1225 | 82 | ||
XM_025155050.1 | XP_025010818.1 | 3 - 4 | 1222 | 82 | ||
GenBank mRNA | AF009012.1 | - | 2 - 3 | 1137 | 82 | |
AY640375.1 | - | 1 - 2 | 880 | 82 | ||
AY687628.1 | - | 1 - 2 | 880 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |