Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204944.2 | NP_990275.1 | 6 - 7 | 2068 | 89 | |
XM_015294136.2 | XP_015149622.1 | 6 - 7 | 2298 | 89 | ||
XM_025155049.1 | XP_025010817.1 | 7 - 8 | 2156 | 89 | ||
XM_025155050.1 | XP_025010818.1 | 7 - 8 | 2153 | 89 | ||
GenBank mRNA | AF009012.1 | - | 6 - 7 | 2068 | 89 | |
AY640375.1 | - | 6 - 7 | 1964 | 89 | ||
AY687628.1 | - | 5 - 6 | 1811 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |