Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205065.1 | NP_990396.1 | 7 - 8 | 1128 | 93 | |
XM_015296828.2 | XP_015152314.1 | 7 - 8 | 1292 | 93 | ||
XM_015296829.2 | XP_015152315.1 | 7 - 8 | 1290 | 93 | ||
XM_015296830.2 | XP_015152316.1 | 7 - 8 | 1291 | 93 | ||
XM_015296831.2 | XP_015152317.1 | 7 - 8 | 1288 | 93 | ||
XM_015296832.2 | XP_015152318.1 | 7 - 8 | 1282 | 93 | ||
XM_025142485.1 | XP_024998253.1 | 7 - 8 | 1281 | 93 | ||
XM_025142486.1 | XP_024998254.1 | 7 - 8 | 1279 | 93 | ||
GenBank mRNA | D87838.1 | - | 7 - 8 | 1128 | 93 | |
DQ471304.1 | - | 6 - 7 | 943 | 93 | ||
KF976429.1 | - | 4 - 5 | 522 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |