Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205175.3 | NP_990506.2 | 5 - 6 | 551 | 80 | |
XM_015275225.2 | XP_015130711.1 | 5 - 6 | 680 | 80 | ||
GenBank mRNA | AY539829.1 | - | 5 - 6 | 551 | 80 | |
AY539830.1 | - | 5 - 6 | 593 | 80 | ||
S52930.1 | - | 4 - 5 | 523 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |