Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205245.1 | NP_990576.1 | 2 - 3 | 297 | 85 | |
GenBank mRNA | BX933363.2 | - | 2 - 3 | 369 | 85 | |
X73533.1 | - | 297 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |