Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205331.1 | NP_990662.1 | 2 - 3 | 582 | 81 | |
GenBank mRNA | X70471.1 | - | 582 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |