Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206904.1 | NP_996787.1 | 3 - 4 | 178 | 72 | |
GenBank mRNA | EU779493.1 | - | 2 - 3 | 67 | 72 | |
Y08916.1 | - | 178 | 72 | |||
Y08917.1 | - | 67 | 72 | |||
Y08918.1 | - | 67 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |